Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpf40a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106480.1 | NP_001099950.1 | 4, 5 | 400 | ||
XM_006234164.2 | XP_006234226.1 | 4, 5 | 725 | |||
XM_008761709.1 | XP_008759931.1 | 4, 5 | 852 | |||
XM_008761710.1 | XP_008759932.1 | 4, 5 | 853 | |||
XM_008761711.1 | XP_008759933.1 | 4, 5 | 725 | |||
XM_008761712.1 | XP_008759934.1 | 4, 5 | 726 | |||
XM_008761713.1 | XP_008759935.1 | 3, 4 | 176 | |||
GenBank mRNA | FQ231481.1 | - | 4, 5 | 319 |