Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Banf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006235132.2 | XP_006235194.1 | 4 | 395 | ||
XM_006235134.2 | XP_006235196.1 | 6 | 1300 | |||
XM_006235135.2 | XP_006235197.1 | 4 | 1089 | |||
XM_006235137.2 | XP_006235199.1 | 3 | 237 | |||
XM_008762281.1 | XP_008760503.1 | 3 | 359 | |||
XM_008762282.1 | XP_008760504.1 | 4 | 1045 | |||
XM_008762283.1 | XP_008760505.1 | 2 | 76 | |||
XM_215865.7 | XP_215865.3 | 3 | 970 |