Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyss | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198685.1 | NP_941958.1 | 3 | 360 | ||
GenBank mRNA | J04206.1 | - | 3 | 334 |