Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100885.1 | NP_001094355.1 | 2, 3 | 359 | ||
XM_006235401.1 | XP_006235463.1 | 3, 4 | 475 | |||
XM_006235402.2 | XP_006235464.1 | 3, 4 | 489 | |||
GenBank mRNA | BC158548.1 | - | 3, 4 | 438 | 9036351 | |
FQ215756.1 | - | 3, 4 | 412 | |||
FQ220245.1 | - | 3, 4 | 416 | |||
FQ220303.1 | - | 3, 4 | 416 | |||
FQ220687.1 | - | 3, 4 | 418 | |||
FQ227515.1 | - | 3, 4 | 419 | |||
FQ229420.1 | - | 3, 4 | 416 | |||
FQ229727.1 | - | 3, 4 | 419 | |||
FQ229762.1 | - | 3, 4 | 418 | |||
FQ229855.1 | - | 3, 4 | 452 | |||
FQ230056.1 | - | 3, 4 | 419 | |||
FQ230407.1 | - | 3, 4 | 432 | |||
FQ231493.1 | - | 3, 4 | 432 | |||
FQ231820.1 | - | 3, 4 | 451 | |||
FQ232091.1 | - | 3, 4 | 412 | |||
S77900.1 | - | 2, 3 | 359 |