Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100885.1 | NP_001094355.1 | 3 | 429 | ||
XM_006235401.1 | XP_006235463.1 | 4 | 545 | |||
XM_006235402.2 | XP_006235464.1 | 4 | 559 | |||
GenBank mRNA | BC158548.1 | - | 4 | 508 | 9036351 | |
FQ215756.1 | - | 4 | 482 | |||
FQ219977.1 | - | 4 | 486 | |||
FQ220245.1 | - | 4 | 486 | |||
FQ220303.1 | - | 4 | 486 | |||
FQ220687.1 | - | 4 | 488 | |||
FQ227515.1 | - | 4 | 489 | |||
FQ229420.1 | - | 4 | 486 | |||
FQ229762.1 | - | 4 | 488 | |||
FQ229855.1 | - | 4 | 522 | |||
FQ230056.1 | - | 4 | 489 | |||
FQ230407.1 | - | 4 | 502 | |||
FQ231493.1 | - | 4 | 502 | |||
FQ231820.1 | - | 4 | 521 | |||
FQ232091.1 | - | 4 | 482 | |||
S77900.1 | - | 3 | 429 |