Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il36a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106554.1 | NP_001100024.1 | 4 | 372 | ||
XM_008761581.1 | XP_008759803.1 | 3 | 993 |