Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110139.2 | NP_001103609.1 | 18 | 3162 | ||
NM_001110823.2 | NP_001104293.1 | 18 | 3162 | |||
GenBank mRNA | AF043106.1 | - | 13 | 2134 | ||
BM387097.1 | - | 3 | 585 | |||
CK358684.1 | - | 3 | 347 | |||
EV765256.1 | - | 4 | 415 | |||
J04022.1 | - | 18 | 3160 | |||
J04023.1 | - | 18 | 2795 | |||
J04024.1 | - | 18 | 2894 | |||
M25267.1 | - | 7 | 1195 | |||
X15635.1 | - | 18 | 2863 |