Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183327.1 | NP_899156.1 | 2 | 194 | ||
XM_006246125.2 | XP_006246187.1 | 2 | 757 | |||
GenBank mRNA | AY574252.1 | - | 2 | 194 |