Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017302.2 | NP_058998.2 | 4 | 592 | ||
XM_006241432.2 | XP_006241494.1 | 5 | 1134 | |||
XM_006241433.2 | XP_006241495.1 | 6 | 2650 | |||
XM_006241434.2 | XP_006241496.1 | 6 | 1001 | |||
XM_006241435.2 | XP_006241497.1 | 5 | 2614 | |||
XM_006241436.2 | XP_006241498.1 | 5 | 876 | |||
XM_006241437.2 | XP_006241499.1 | 5 | 2524 | |||
GenBank mRNA | BC081701.1 | - | 4 | 592 | 7130138 | |
FQ218176.1 | - | 5 | 706 | |||
U62316.1 | - | 5 | 758 | |||
X97445.1 | - | 5 | 717 |