Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034131.1 | NP_001029303.1 | 8 | 1145 | ||
XM_006236954.2 | XP_006237016.1 | 7 | 871 | |||
XM_006236955.2 | XP_006237017.1 | 7 | 1386 | |||
XM_006236956.1 | XP_006237018.1 | 7 | 808 | |||
XM_008763148.1 | XP_008761370.1 | 10 | 1887 | |||
XM_008763149.1 | XP_008761371.1 | 10 | 1289 | |||
XM_008763150.1 | XP_008761372.1 | 11 | 1364 | |||
XM_008763151.1 | XP_008761373.1 | 10 | 1481 | |||
XM_008763152.1 | XP_008761374.1 | 9 | 1467 | |||
XM_008763153.1 | XP_008761375.1 | 9 | 2013 | |||
XM_008763154.1 | XP_008761376.1 | 10 | 1884 | |||
XM_008763155.1 | XP_008761377.1 | 10 | 1885 | |||
XM_008763156.1 | XP_008761378.1 | 10 | 1888 | |||
XM_008763157.1 | XP_008761379.1 | 10 | 1286 | |||
XM_008763158.1 | XP_008761380.1 | 10 | 1289 | |||
GenBank mRNA | BC100267.1 | - | 8 | 1145 | 7383433 |