Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpp25l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106648.2 | NP_001100118.1 | 2 | 295 | ||
XM_006238031.2 | XP_006238093.1 | 2 | 463 | |||
XM_006238032.2 | XP_006238094.1 | 2 | 459 | |||
GenBank mRNA | BC158601.1 | - | 2 | 295 | 7133091 |