Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhox9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024874.1 | NP_001020045.1 | 2 | 213 | ||
GenBank mRNA | DQ058656.1 | - | 2 | 213 |
Gene Symbol: LOC100364002 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257483.2 | XP_006257545.1 | 2 | 205 |