Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pink1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106694.1 | NP_001100164.1 | 1, 2 | 414 | ||
GenBank mRNA | BC169047.1 | - | 1, 2 | 408 | 7190543 |