Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrto4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106697.1 | NP_001100167.1 | 7, 8 | 1000 | ||
XM_006239178.2 | XP_006239240.1 | 7, 8 | 868 | |||
XM_008764230.1 | XP_008762452.1 | 6, 7 | 602 | |||
GenBank mRNA | BC168898.1 | - | 7, 8 | 567 | ||
FQ227354.1 | - | 7, 8 | 613 | |||
FQ227472.1 | - | 7, 8 | 852 | |||
FQ234412.1 | - | 7, 8 | 852 |