Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bok | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017312.2 | NP_059008.1 | 4, 5 | 611 | ||
GenBank mRNA | AF027954.1 | - | 3, 4 | 521 | ||
AF051093.1 | - | 2, 3 | 392 | |||
BC078871.1 | - | 4, 5 | 611 | 7132448 | ||
FQ227820.1 | - | 3, 4 | 452 |