Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1310769 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106749.1 | NP_001100219.1 | 3, 4 | 719 | ||
XM_006240367.2 | XP_006240429.1 | 2, 3 | 528 | |||
XM_006240368.1 | XP_006240430.1 | 2, 3 | 247 | |||
GenBank mRNA | FQ212891.1 | - | 2, 3 | 207 |