Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134529.1 | NP_001128001.1 | 3 | 386 | ||
XM_006241976.2 | XP_006242038.1 | 3 | 532 | |||
XM_006241977.2 | XP_006242039.1 | 3 | 526 | |||
XM_006241978.2 | XP_006242040.1 | 3 | 529 | |||
XM_006241979.2 | XP_006242041.1 | 3 | 526 | |||
XM_006241981.2 | XP_006242043.1 | 3 | 523 | |||
XM_006241982.2 | XP_006242044.1 | 3 | 522 | |||
XM_006241983.2 | XP_006242045.1 | 3 | 522 | |||
XM_008765708.1 | XP_008763930.1 | 3 | 529 | |||
XM_008765709.1 | XP_008763931.1 | 3 | 527 | |||
XM_008765710.1 | XP_008763932.1 | 3 | 530 |