Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbd3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106803.1 | NP_001100273.1 | 3 | 260 | ||
XM_006242571.2 | XP_006242633.1 | 2 | 345 | |||
XM_006242574.2 | XP_006242636.1 | 3 | 621 | |||
XM_006242576.2 | XP_006242638.1 | 2 | 176 |