Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1240 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000448.1 | NP_001000448.1 | 1 | 436 |
Gene Symbol: LOC103692019 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762664.1 | XP_008760886.1 | 1 | 436 |