Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1259 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000455.1 | NP_001000455.1 | 1 | 600 |
Gene Symbol: LOC100912340 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749617.3 | XP_003749665.2 | 1 | 698 |