Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1273 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000458.1 | NP_001000458.1 | 658 |
Gene Symbol: LOC100909937 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749259.1 | XP_003749307.1 | 1 | 658 |
Gene Symbol: LOC100909958 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749180.1 | XP_003749228.1 | 658 |
Gene Symbol: LOC100910086 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749262.1 | XP_003749310.1 | 658 |
Gene Symbol: LOC100910153 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750490.1 | XP_003750538.1 | 1 | 658 |
Gene Symbol: LOC100910227 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749265.1 | XP_003749313.1 | 658 |
Gene Symbol: LOC100910363 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749186.1 | XP_003749234.1 | 1 | 658 |
Gene Symbol: LOC100910548 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750497.1 | XP_003750545.1 | 1 | 658 |