Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024762.1 | NP_001019933.1 | 6, 7 | 670 | ||
XM_006244686.2 | XP_006244748.1 | 4, 5 | 499 | |||
XM_006244689.2 | XP_006244751.1 | 4, 5 | 450 | |||
XM_006244690.2 | XP_006244752.1 | 4, 5 | 516 | |||
XM_006244691.1 | XP_006244753.1 | 4, 5 | 517 | |||
GenBank mRNA | AJ011607.1 | - | 3, 4 | 354 | ||
BC097454.1 | - | 6, 7 | 670 | 7456950 |