Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glb1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127529.2 | NP_001121001.1 | 16 | 1898 | ||
XM_006245177.2 | XP_006245239.1 | 16 | 2056 | |||
XM_006245179.2 | XP_006245241.1 | 16 | 1849 | |||
XM_006245180.2 | XP_006245242.1 | 16 | 1861 | |||
XM_006245182.2 | XP_006245244.1 | 16 | 1747 | |||
XM_008767223.1 | XP_008765445.1 | 15 | 2954 | |||
XM_008767224.1 | XP_008765446.1 | 16 | 2471 | |||
XM_008767225.1 | XP_008765447.1 | 13 | 2684 | |||
GenBank mRNA | BC161869.1 | - | 16 | 1898 | 7109779 |