Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif1ax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106963.1 | NP_001100433.1 | 4, 5 | 476 | ||
GenBank mRNA | BC168938.1 | - | 4, 5 | 349 | 7097069 | |
FQ217925.1 | - | 4, 5 | 441 | |||
FQ221854.1 | - | 4, 5 | 455 | |||
FQ222873.1 | - | 4, 5 | 474 |