Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEWGENE_1359295 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767502.1 | XP_008765724.1 | 1069 |
Gene Symbol: Rsl1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008876.2 | NP_001008876.1 | 6 | 496 | ||
GenBank mRNA | AY310144.1 | - | 6 | 496 |