Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubald1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007668.1 | NP_001007669.1 | 2, 3 | 228 | ||
GenBank mRNA | BC079459.1 | - | 2, 3 | 228 | 7121902 | |
FQ214502.1 | - | 2, 3 | 265 | |||
FQ228825.1 | - | 2, 3 | 256 |
Gene Symbol: LOC100910875 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750772.3 | XP_003750820.1 | 926 |