Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neurl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107013.1 | NP_001100483.1 | 25 | 3992 | ||
XM_006246712.1 | XP_006246774.1 | 25 | 3992 | |||
XM_008767829.1 | XP_008766051.1 | 23 | 3411 |