Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107021.1 | NP_001100491.1 | 622 |
Gene Symbol: LOC100912068 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750862.3 | XP_003750910.2 | 2 | 427 |