Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182819.2 | NP_877971.2 | 3 | 242 | ||
GenBank mRNA | AY339885.1 | - | 3 | 202 | ||
BC079123.1 | - | 3 | 242 | 7126762 | ||
FQ210062.1 | - | 3 | 237 | |||
FQ213156.1 | - | 3 | 273 | |||
FQ214913.1 | - | 3 | 272 | |||
FQ217174.1 | - | 3 | 267 | |||
FQ217253.1 | - | 3 | 260 | |||
FQ217570.1 | - | 3 | 273 | |||
FQ217933.1 | - | 3 | 250 | |||
FQ217995.1 | - | 3 | 272 | |||
FQ218015.1 | - | 3 | 272 | |||
FQ218434.1 | - | 3 | 272 | |||
FQ221598.1 | - | 3 | 272 | |||
FQ221667.1 | - | 3 | 271 | |||
FQ222019.1 | - | 3 | 271 | |||
FQ222086.1 | - | 3 | 260 | |||
FQ222256.1 | - | 3 | 272 | |||
FQ222576.1 | - | 3 | 264 | |||
FQ223858.1 | - | 3 | 260 | |||
FQ224255.1 | - | 3 | 260 | |||
FQ224424.1 | - | 3 | 272 | |||
FQ224448.1 | - | 3 | 272 | |||
FQ225820.1 | - | 3 | 272 | |||
FQ229714.1 | - | 3 | 273 |