Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sirt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107073.1 | NP_001100543.1 | 6 | 575 | ||
XM_006247889.2 | XP_006247951.1 | 6 | 624 | |||
XM_008768477.1 | XP_008766699.1 | 6 | 624 | |||
GenBank mRNA | BC167031.1 | - | 6 | 557 | 7099085 |