Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgcr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107080.2 | NP_001100550.1 | 5 | 549 | ||
XM_006248609.2 | XP_006248671.1 | 5 | 587 | |||
GenBank mRNA | BC158828.1 | - | 5 | 549 | 7378165 |