Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc51a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107087.1 | NP_001100557.1 | 3 | 510 | ||
XM_008768725.1 | XP_008766947.1 | 3 | 331 |