Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195558.1 | NP_001182487.1 | 1, 2 | 538 | ||
GenBank mRNA | CB547021.1 | - | 1, 2 | 251 |