Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195558.1 | NP_001182487.1 | 2 | 597 | ||
GenBank mRNA | CB547021.1 | - | 2 | 310 |