Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oas1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009379.1 | NP_001009379.1 | 485 | |||
GenBank mRNA | AY196699.1 | - | 485 |
Gene Symbol: LOC103689988 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769299.1 | XP_008767521.1 | 2, 3 | 585 |