Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007677.1 | NP_001007678.1 | 2 | 389 | ||
GenBank mRNA | BC081864.1 | - | 2 | 389 | 7096152 |