Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfyve28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107227.1 | NP_001100697.1 | 11 | 2592 | ||
XM_006251256.2 | XP_006251318.1 | 12 | 2718 | |||
XM_006251257.2 | XP_006251319.1 | 12 | 2718 |