Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107273.1 | NP_001100743.1 | 10 | 1325 | ||
XM_006252117.2 | XP_006252179.1 | 10 | 1399 | |||
XM_006252118.2 | XP_006252180.1 | 10 | 1507 | |||
XM_006252120.2 | XP_006252182.1 | 10 | 1396 | |||
XM_006252121.2 | XP_006252183.1 | 7 | 1453 | |||
XM_008770757.1 | XP_008768979.1 | 7 | 779 | |||
GenBank mRNA | BC087063.1 | - | 10 | 1352 | ||
BC105847.1 | - | 10 | 1395 | |||
FQ211026.1 | - | 10 | 1306 |