Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107281.1 | NP_001100751.1 | 4 | 407 | ||
XM_006252408.2 | XP_006252470.1 | 4 | 409 | |||
XM_006252409.2 | XP_006252471.1 | 4 | 404 | |||
XM_006252410.2 | XP_006252472.1 | 4 | 475 | |||
XM_006252413.2 | XP_006252475.1 | 3 | 257 | |||
XM_006252415.2 | XP_006252477.1 | 3 | 254 | |||
XM_008770870.1 | XP_008769092.1 | 3 | 2211 | |||
XM_008770871.1 | XP_008769093.1 | 5 | 421 |