Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipo5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252478.2 | XP_006252540.1 | 23 | 2889 | ||
XM_224534.8 | XP_224534.7 | 24 | 3135 | |||
GenBank mRNA | FQ214398.1 | - | 3 | 286 |