Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pstpip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271281.1 | NP_001258210.1 | 11, 12 | 1025 | ||
XM_008772138.1 | XP_008770360.1 | 10, 11 | 1278 | |||
XM_008772139.1 | XP_008770361.1 | 10, 11 | 1278 | |||
GenBank mRNA | CK364772.1 | - | 3, 4 | 250 |