Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107467.2 | NP_001100937.1 | 4 | 1514 | ||
XM_006228023.2 | XP_006228085.1 | 4 | 1879 | |||
XM_006228024.2 | XP_006228086.1 | 4 | 1723 | |||
XM_006228025.1 | XP_006228087.1 | 4 | 1483 | |||
XM_006228026.2 | XP_006228088.1 | 4 | 1452 | |||
GenBank mRNA | BC168865.1 | - | 4 | 1514 | 7134370 |