Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pvrl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012064.1 | NP_001012064.1 | 6, 7 | 1390 | ||
GenBank mRNA | BC090075.1 | - | 6, 7 | 1390 | 7189358 |