Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hddc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107528.1 | NP_001100998.1 | 1, 2 | 135 | ||
XM_008759531.1 | XP_008757753.1 | 1, 2 | 158 |