Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdpgp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229451.2 | XP_006229513.1 | 2 | 807 | ||
XM_008759586.1 | XP_008757808.1 | 3 | 1160 | |||
XM_218822.7 | XP_218822.2 | 3 | 936 |