Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ovol1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107572.1 | NP_001101042.1 | 1 | 349 | ||
XM_006230794.2 | XP_006230856.1 | 1 | 2358 | |||
XM_006230796.2 | XP_006230858.1 | 1 | 22 |