Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107596.1 | NP_001101066.1 | 9, 10 | 2057 | ||
XM_006231163.1 | XP_006231225.1 | 9, 10 | 2070 | |||
XM_006231164.2 | XP_006231226.1 | 10, 11 | 2344 | |||
XM_006231165.2 | XP_006231227.1 | 10, 11 | 3182 | |||
XM_006231166.2 | XP_006231228.1 | 10, 11 | 2178 | |||
XM_008760298.1 | XP_008758520.1 | 11, 12 | 2519 | |||
XM_008760299.1 | XP_008758521.1 | 10, 11 | 2230 |