Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neurl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107605.1 | NP_001101075.1 | 2 | 376 | ||
XM_006231510.2 | XP_006231572.1 | 2 | 490 | |||
XM_006231511.2 | XP_006231573.1 | 2 | 347 | |||
XM_006231512.2 | XP_006231574.1 | 2 | 890 |