Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myof | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191636.1 | NP_001178565.1 | 29 | 3308 | ||
XM_006231325.2 | XP_006231387.1 | 30 | 3676 | |||
XM_006231326.2 | XP_006231388.1 | 29 | 3637 | |||
XM_008760368.1 | XP_008758590.1 | 30 | 3676 | |||
XM_008760369.1 | XP_008758591.1 | 30 | 3676 |