Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epb41l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006227709.1 | XP_006227771.1 | 3, 4 | 873 | ||
XM_008758646.1 | XP_008756868.1 | 3, 4 | 1055 | |||
XM_008758647.1 | XP_008756869.1 | 3, 4 | 1171 | |||
XM_008758648.1 | XP_008756870.1 | 3, 4 | 776 | |||
XM_008758649.1 | XP_008756871.1 | 4, 5 | 1011 | |||
XM_008758650.1 | XP_008756872.1 | 3, 4 | 3858 | |||
GenBank mRNA | FQ212032.1 | - | 4, 5 | 969 |